Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024285.1 | NP_001019456.1 | 11 | 2054 | ||
GenBank mRNA | AB174900.2 | - | 11 | 2054 | ||
AY367561.1 | - | 11 | 2054 |