Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADGRG1 SIRNA Assays ›
Gene Symbol:
ADGRG1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
BFPP, BPPR, GPR56, TM7LN4, TM7XN1
Chromosome Location:
Chr.16: 57619535 - 57665039 on Build GRCh38
UniGene ID:
Hs.513633
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17757
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADGRG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145770.2
NP_001139242.1
4
585
NM_001145771.2
NP_001139243.1
4
1023
NM_001145772.2
NP_001139244.1
4
1049
NM_001145773.2
NP_001139245.1
3
920
NM_001145774.2
NP_001139246.1
4
822
NM_001290143.1
NP_001277072.1
3
474
NM_001290144.1
NP_001277073.1
3
661
NM_005682.6
NP_005673.3
4
585
NM_201524.3
NP_958932.1
4
1027
NM_201525.3
NP_958933.1
3
905
XM_005256237.2
XP_005256294.1
3
621
XM_005256238.2
XP_005256295.1
4
960
XM_005256239.2
XP_005256296.1
4
722
XM_005256240.4
XP_005256297.1
3
443
XM_005256241.4
XP_005256298.1
3
447
XM_005256242.4
XP_005256299.1
3
504
XM_005256244.3
XP_005256301.1
3
596
XM_005256245.2
XP_005256302.1
3
647
XM_005256246.2
XP_005256303.1
4
707
XM_005256247.2
XP_005256304.1
3
606
XM_005256248.2
XP_005256305.1
3
671
XM_005256249.3
XP_005256306.1
3
462
XM_005256251.4
XP_005256308.1
3
488
XM_005256252.2
XP_005256309.1
3
432
XM_005256254.2
XP_005256311.1
3
632
XM_005256255.2
XP_005256312.1
4
945
XM_006721338.2
XP_006721401.1
4
743
XM_006721339.3
XP_006721402.1
4
550
XM_006721340.3
XP_006721403.1
4
808
XM_006721341.2
XP_006721404.1
4
765
XM_006721342.2
XP_006721405.1
4
728
XM_006721343.3
XP_006721406.1
4
793
XM_006721344.2
XP_006721407.1
4
750
XM_006721345.3
XP_006721408.1
4
519
XM_006721346.2
XP_006721409.1
3
374
XM_006721347.2
XP_006721410.1
4
960
XM_011523461.2
XP_011521763.1
4
711
XM_011523462.2
XP_011521764.1
3
526
XM_011523463.2
XP_011521765.1
4
536
XM_011523464.2
XP_011521766.1
4
696
XM_011523465.2
XP_011521767.1
4
634
XM_011523466.2
XP_011521768.1
3
681
XM_011523467.2
XP_011521769.1
4
428
XM_011523468.2
XP_011521770.1
3
430
XM_017023892.1
XP_016879381.1
3
632
GenBank mRNA
AF106858.1
-
3
455
AJ011001.1
-
4
609
AK075270.1
-
4
537
AK075555.1
-
4
560
AK297236.1
-
3
534
AK299110.1
-
3
449
AK312405.1
-
3
741
AY358400.1
-
4
506
BC008770.2
-
3
451
3139174
BC013207.1
-
4
599
BE893273.1
-
3
510
BT007311.1
-
2
293
CR936747.1
-
3
416
DA386131.1
-
3
467
EU432119.1
-
2
293
KU178472.1
-
2
293
KU178473.1
-
2
293
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Matching Taqman Gene Expression Assay:
Hs00173754_m1
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
cell adhesion
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
Rho protein signal transduction
cell-cell signaling
brain development
negative regulation of cell proliferation
vascular endothelial growth factor production
cell migration
cerebral cortex regionalization
cerebral cortex radial glia guided migration
layer formation in cerebral cortex
positive regulation of Rho protein signal transduction
positive regulation of cell adhesion
protein kinase C signaling
positive regulation of neural precursor cell proliferation
negative regulation of neuron migration
G-protein coupled receptor activity
collagen binding
heparin binding
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline