Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADGRG1 SIRNA Assays ›
Gene Symbol:
ADGRG1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
BFPP, BPPR, GPR56, TM7LN4, TM7XN1
Chromosome Location:
Chr.16: 57619535 - 57665039 on Build GRCh38
UniGene ID:
Hs.513633
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17758
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADGRG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145770.2
NP_001139242.1
6
931
NM_001145771.2
NP_001139243.1
6
1369
NM_001145772.2
NP_001139244.1
6
1395
NM_001145773.2
NP_001139245.1
5
1266
NM_001145774.2
NP_001139246.1
6
1168
NM_001290142.1
NP_001277071.1
5
471
NM_001290143.1
NP_001277072.1
5
820
NM_001290144.1
NP_001277073.1
5
1007
NM_005682.6
NP_005673.3
6
931
NM_201524.3
NP_958932.1
6
1373
NM_201525.3
NP_958933.1
5
1251
XM_005256237.2
XP_005256294.1
5
967
XM_005256238.2
XP_005256295.1
6
1306
XM_005256239.2
XP_005256296.1
6
1068
XM_005256240.4
XP_005256297.1
5
789
XM_005256241.4
XP_005256298.1
5
793
XM_005256242.4
XP_005256299.1
5
850
XM_005256244.3
XP_005256301.1
5
942
XM_005256245.2
XP_005256302.1
5
993
XM_005256246.2
XP_005256303.1
6
1053
XM_005256247.2
XP_005256304.1
5
952
XM_005256248.2
XP_005256305.1
5
1017
XM_005256249.3
XP_005256306.1
5
808
XM_005256251.4
XP_005256308.1
5
834
XM_005256252.2
XP_005256309.1
5
778
XM_005256254.2
XP_005256311.1
5
978
XM_005256255.2
XP_005256312.1
6
1291
XM_006721338.2
XP_006721401.1
6
1089
XM_006721339.3
XP_006721402.1
6
896
XM_006721340.3
XP_006721403.1
6
1154
XM_006721341.2
XP_006721404.1
6
1111
XM_006721342.2
XP_006721405.1
6
1074
XM_006721343.3
XP_006721406.1
6
1139
XM_006721344.2
XP_006721407.1
6
1096
XM_006721345.3
XP_006721408.1
6
865
XM_006721346.2
XP_006721409.1
5
720
XM_006721347.2
XP_006721410.1
6
1306
XM_011523461.2
XP_011521763.1
6
1057
XM_011523462.2
XP_011521764.1
5
872
XM_011523463.2
XP_011521765.1
6
882
XM_011523464.2
XP_011521766.1
6
1042
XM_011523465.2
XP_011521767.1
6
980
XM_011523466.2
XP_011521768.1
5
1027
XM_011523467.2
XP_011521769.1
6
774
XM_011523468.2
XP_011521770.1
5
776
XM_017023892.1
XP_016879381.1
5
978
GenBank mRNA
AF106858.1
-
5
801
AJ011001.1
-
6
955
AK075270.1
-
6
883
AK075555.1
-
6
906
AK131550.1
-
5
426
AK297236.1
-
5
880
AK299110.1
-
5
795
AK312405.1
-
5
1087
AY358400.1
-
6
852
BC008770.2
-
5
797
3139174
BC013207.1
-
6
945
BT007311.1
-
4
639
CR936747.1
-
5
762
EU432119.1
-
4
639
KU178472.1
-
4
639
KU178473.1
-
4
639
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Matching Taqman Gene Expression Assay:
Hs00173754_m1
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
cell adhesion
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
Rho protein signal transduction
cell-cell signaling
brain development
negative regulation of cell proliferation
vascular endothelial growth factor production
cell migration
cerebral cortex regionalization
cerebral cortex radial glia guided migration
layer formation in cerebral cortex
positive regulation of Rho protein signal transduction
positive regulation of cell adhesion
protein kinase C signaling
positive regulation of neural precursor cell proliferation
negative regulation of neuron migration
G-protein coupled receptor activity
collagen binding
heparin binding
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline