Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005684.4 | NP_005675.3 | 1 | 477 | ||
GenBank mRNA | BC067456.1 | - | 1 | 439 | 6971753 | |
BC067457.1 | - | 1 | 439 | 6971754 | ||
BC067458.1 | - | 1 | 439 | 6971755 | ||
BC067459.1 | - | 1 | 439 | 6971756 | ||
BC067460.1 | - | 1 | 439 | 6971757 | ||
BC069460.1 | - | 1 | 439 | 7262282 | ||
BC095532.1 | - | 1 | 478 | 7002024 | ||
HQ995526.1 | - | 1 | 439 |
Set Membership: |
G-protein coupled receptor |