Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lilrb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228238.2 | XP_006228300.1 | 1 | 71 | ||
XM_006228239.2 | XP_006228301.1 | 1 | 71 |