Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1565655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177686.1 | NP_001171157.2 | 5 | 782 | ||
XM_008758968.1 | XP_008757190.1 | 503 | ||||
GenBank mRNA | BC158626.1 | - | 5 | 782 | 7455240 |
Gene Symbol: LOC102553892 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759075.1 | XP_008757297.1 | 4 | 503 | ||
XM_008759076.1 | XP_008757298.1 | 4 | 503 |
Gene Symbol: RGD1564380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758911.1 | XP_008757133.1 | 5 | 828 | ||
XM_008758912.1 | XP_008757134.1 | 5 | 836 | |||
XM_008758913.1 | XP_008757135.1 | 5 | 689 | |||
XM_008758914.1 | XP_008757136.1 | 5 | 831 | |||
XM_008758915.1 | XP_008757137.1 | 5 | 828 |