Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004231.3 | NP_004222.2 | 1, 2 | 254 | ||
GenBank mRNA | AK312214.1 | - | 1, 2 | 199 | ||
BC104230.1 | - | 1, 2 | 205 | 40032234 | ||
BC104231.1 | - | 1, 2 | 205 | 40032235 | ||
BC107854.1 | - | 1, 2 | 231 | 5171984 | ||
BG724326.1 | - | 1, 2 | 239 | |||
CR456896.1 | - | 1, 2 | 152 | |||
D49400.1 | - | 1, 2 | 202 | |||
DB499437.1 | - | 1, 2 | 254 |