Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024286.1 | NP_001019457.1 | 3 | 102 | ||
XM_006229111.2 | XP_006229173.1 | 2 | 851 | |||
XM_006229112.2 | XP_006229174.1 | 3 | 171 | |||
XM_006229113.2 | XP_006229175.1 | 2 | 851 | |||
XM_006229114.2 | XP_006229176.1 | 2 | 796 | |||
XM_006229115.2 | XP_006229177.1 | 3 | 116 | |||
XM_008759404.1 | XP_008757626.1 | 2 | 799 | |||
XM_008759405.1 | XP_008757627.1 | 2 | 152 | |||
GenBank mRNA | BC089881.1 | - | 3 | 102 | 7368473 |