Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scube2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748970.3 | XP_003749018.1 | 19 | 2525 | ||
XM_006230005.2 | XP_006230067.1 | 18 | 2402 | |||
XM_006230006.2 | XP_006230068.1 | 15 | 2063 |