Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Knop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134611.2 | NP_001128083.1 | 2 | 437 | ||
XM_008759787.1 | XP_008758009.1 | 2 | 445 | |||
XM_008759788.1 | XP_008758010.1 | 2 | 445 | |||
XM_008759789.1 | XP_008758011.1 | 2 | 578 |