Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprcap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024289.1 | NP_001019460.1 | 2 | 804 | ||
GenBank mRNA | BC088279.1 | - | 2 | 804 | 7382668 |