Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314052.1 | NP_001300981.1 | 4 | 1986 | ||
NM_004235.5 | NP_004226.3 | 5 | 1884 | |||
GenBank mRNA | AF022184.1 | - | 5 | 1735 | ||
AF105036.1 | - | 5 | 1638 | |||
AK095134.1 | - | 3 | 1390 | |||
AK095894.1 | - | 1 | 1133 | |||
AK313489.1 | - | 5 | 1473 | |||
BC029923.1 | - | 5 | 1449 | 5111134 | ||
BC030811.1 | - | 3 | 1440 | 4703002 | ||
HF546201.1 | - | 4 | 592 | |||
HF546202.1 | - | 3 | 427 | |||
HF546213.1 | - | 4 | 592 | |||
U70663.1 | - | 5 | 1812 |