Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314052.1 | NP_001300981.1 | 3 | 999 | ||
NM_004235.5 | NP_004226.3 | 3 | 999 | |||
GenBank mRNA | AF022184.1 | - | 3 | 850 | ||
AF105036.1 | - | 3 | 753 | |||
AK095134.1 | - | 1 | 505 | |||
AK313489.1 | - | 3 | 588 | |||
BC029923.1 | - | 3 | 564 | 5111134 | ||
BC030811.1 | - | 2 | 453 | 4703002 | ||
U70663.1 | - | 3 | 927 |