Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100910973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FM045317.1 | - | 2 | 91 | ||
FM097063.1 | - | 2 | 109 | |||
FM098643.1 | - | 2 | 281 | |||
FM110991.1 | - | 2 | 84 | |||
FQ212263.1 | - | 2 | 105 | |||
FQ230677.1 | - | 2 | 105 | |||
FQ230710.1 | - | 2 | 105 | |||
FQ231880.1 | - | 2 | 103 | |||
FQ234947.1 | - | 2 | 101 |