Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037793.2 | NP_001032882.2 | 22, 23 | 2901 | ||
XM_006231244.2 | XP_006231306.1 | 22, 23 | 3112 | |||
XM_006231245.2 | XP_006231307.1 | 21, 22 | 3676 | |||
XM_008760310.1 | XP_008758532.1 | 23, 24 | 3167 | |||
XM_008760311.1 | XP_008758533.1 | 22, 23 | 2753 |