Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047108.1 | NP_001040573.1 | 2 | 377 | ||
GenBank mRNA | AB197923.1 | - | 2 | 377 |