Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034916.2 | NP_001030088.2 | 264 | |||
GenBank mRNA | BC126063.1 | - | 254 | 7307697 | ||
CF111102.1 | - | 264 | ||||
FQ217739.1 | - | 263 | ||||
FQ225104.1 | - | 264 | ||||
FQ226443.1 | - | 262 | ||||
FQ226648.1 | - | 262 | ||||
FQ230224.1 | - | 265 | ||||
FQ230609.1 | - | 271 | ||||
FQ231472.1 | - | 265 | ||||
FQ234329.1 | - | 271 |
Gene Symbol: LOC100360117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729143.4 | XP_002729189.1 | 1 | 277 |
Gene Symbol: LOC100910370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750368.3 | XP_003750416.1 | 3 | 343 |