Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025144.1 | NP_001020315.1 | 7 | 950 | ||
GenBank mRNA | BC079112.1 | - | 7 | 950 | 7125748 |