Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIP12 SIRNA Assays ›
Gene Symbol:
TRIP12
Gene Name:
thyroid hormone receptor interactor 12
Gene Aliases:
TRIP-12, ULF
Chromosome Location:
Chr.2: 229763837 - 229923234 on Build GRCh38
UniGene ID:
Hs.572642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17810
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIP12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284214.1
NP_001271143.1
39
5839
NM_001284215.1
NP_001271144.1
37
5664
NM_001284216.1
NP_001271145.1
36
4885
NM_004238.2
NP_004229.1
38
5695
XM_005246954.4
XP_005247011.1
39
5947
XM_005246955.4
XP_005247012.1
39
5946
XM_005246956.4
XP_005247013.1
39
5923
XM_005246957.4
XP_005247014.1
39
5898
XM_005246958.4
XP_005247015.1
39
5866
XM_005246960.4
XP_005247017.1
40
5860
XM_005246961.3
XP_005247018.1
38
5797
XM_005246962.4
XP_005247019.1
38
5794
XM_006712852.3
XP_006712915.1
38
5750
XM_011512180.2
XP_011510482.1
39
5900
XM_011512181.2
XP_011510483.1
39
5898
XM_011512183.2
XP_011510485.1
38
5754
XM_017005272.1
XP_016860761.1
40
6006
XM_017005273.1
XP_016860762.1
39
5873
XM_017005274.1
XP_016860763.1
39
5821
XM_017005275.1
XP_016860764.1
39
5876
XM_017005276.1
XP_016860765.1
39
5881
XM_017005277.1
XP_016860766.1
40
6021
XM_017005278.1
XP_016860767.1
39
5836
XM_017005279.1
XP_016860768.1
39
5893
XM_017005280.1
XP_016860769.1
39
5882
XM_017005281.1
XP_016860770.1
39
5816
XM_017005282.1
XP_016860771.1
40
5859
XM_017005283.1
XP_016860772.1
39
5848
XM_017005284.1
XP_016860773.1
40
5842
XM_017005285.1
XP_016860774.1
38
5784
XM_017005286.1
XP_016860775.1
38
5816
XM_017005287.1
XP_016860776.1
38
5782
XM_017005288.1
XP_016860777.1
38
5744
XM_017005289.1
XP_016860778.1
40
5779
XM_017005290.1
XP_016860779.1
40
5776
XM_017005291.1
XP_016860780.1
39
5710
XM_017005292.1
XP_016860781.1
38
5698
XM_017005293.1
XP_016860782.1
38
5713
GenBank mRNA
BC113891.1
-
36
4779
40083155
BC114556.1
-
39
5733
40083165
D28476.1
-
38
5625
EU489742.1
-
37
5618
L40383.1
-
2
116
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
DNA repair
cellular response to DNA damage stimulus
embryo development
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of histone H2A K63-linked ubiquitination
negative regulation of double-strand break repair
ubiquitin-protein transferase activity
protein binding
ligase activity
thyroid hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-s7sbm:80/100.66.79.121:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline