Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288962.1 | NP_001275891.1 | 11 | 1309 | ||
NM_001288963.1 | NP_001275892.1 | 10 | 1141 | |||
NM_004240.3 | NP_004231.1 | 10 | 1141 | |||
NR_110231.1 | 11 | 1160 | ||||
XM_006722940.1 | XP_006723003.1 | 11 | 1309 | |||
GenBank mRNA | AB007845.1 | - | 10 | 1116 | ||
AB072596.1 | - | 11 | 1135 | |||
AF380114.1 | - | 10 | 1061 | |||
AF502289.1 | - | 11 | 1268 | |||
AJ000414.1 | - | 10 | 1100 | |||
AK223109.1 | - | 10 | 1106 | |||
AK313296.1 | - | 10 | 1123 | |||
AL832186.1 | - | 11 | 1161 | |||
AY081141.1 | - | 10 | 1095 | |||
BC013002.2 | - | 10 | 1086 | 3532036 | ||
BT006698.1 | - | 10 | 1061 | |||
BT020167.1 | - | 10 | 1061 | |||
BT020171.1 | - | 10 | 1061 | |||
CR536513.1 | - | 10 | 1061 |