Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288962.1 | NP_001275891.1 | 4 | 364 | ||
NM_001288963.1 | NP_001275892.1 | 4 | 364 | |||
NM_004240.3 | NP_004231.1 | 4 | 364 | |||
NR_110231.1 | 4 | 364 | ||||
XM_005259683.2 | XP_005259740.1 | 4 | 364 | |||
XM_006722940.1 | XP_006723003.1 | 4 | 364 | |||
GenBank mRNA | AB007845.1 | - | 4 | 339 | ||
AB072596.1 | - | 4 | 339 | |||
AF380114.1 | - | 4 | 284 | |||
AF502289.1 | - | 4 | 323 | |||
AJ000414.1 | - | 4 | 323 | |||
AK223109.1 | - | 4 | 329 | |||
AK313296.1 | - | 4 | 346 | |||
AL832186.1 | - | 4 | 450 | |||
AY081141.1 | - | 4 | 318 | |||
BC013002.2 | - | 4 | 309 | 3532036 | ||
BG494851.1 | - | 368 | ||||
BT006698.1 | - | 4 | 284 | |||
BT020167.1 | - | 4 | 284 | |||
BT020171.1 | - | 4 | 284 | |||
CB997985.1 | - | 4 | 377 | |||
CR536513.1 | - | 4 | 284 | |||
DB198268.1 | - | 4 | 366 |