Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109177.1 | NP_001102647.1 | 13 | 1865 | ||
XM_006232024.2 | XP_006232086.1 | 13 | 1856 |