Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNHIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281433.1 | NP_001268362.1 | 5 | 355 | ||
NM_001281434.1 | NP_001268363.1 | 3 | 221 | |||
NM_004773.3 | NP_004764.1 | 5 | 389 | |||
NR_104009.1 | 5 | 384 | ||||
NR_104010.1 | 4 | 357 | ||||
GenBank mRNA | AB209193.1 | - | 3 | 3853 | ||
AF400652.1 | - | 5 | 381 | |||
AK290858.1 | - | 5 | 340 | |||
BC017931.1 | - | 5 | 354 | 4669658 | ||
BC105608.1 | - | 4 | 329 | |||
BE260293.1 | - | 3 | 120 | |||
BG032116.1 | - | 5 | 290 | |||
BM457955.1 | - | 319 | ||||
CB996852.1 | - | 5 | 415 | |||
HY045632.1 | - | 5 | 390 | |||
L40410.1 | - | 5 | 307 |