Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfxap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044263.1 | NP_001037728.1 | 734 | |||
GenBank mRNA | BC098816.1 | - | 734 | 7377196 |
Gene Symbol: Rfxapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761021.1 | XP_008759243.1 | 3 | 761 |