Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1562234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109187.1 | NP_001102657.1 | 2 | 171 | ||
XM_006232838.2 | XP_006232900.1 | 3 | 279 |