Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004775.3 | NP_004766.2 | 2, 3 | 530 | ||
XM_005258387.4 | XP_005258444.1 | 2, 3 | 295 | |||
XM_006722579.3 | XP_006722642.1 | 3, 4 | 243 | |||
XM_017026089.1 | XP_016881578.1 | 2, 3 | 701 | |||
XM_017026092.1 | XP_016881581.1 | 2, 3 | 701 | |||
GenBank mRNA | AB024742.2 | - | 2, 3 | 239 | ||
AF038664.1 | - | 2, 3 | 508 | |||
AF097159.1 | - | 2, 3 | 530 | |||
AK314860.1 | - | 2, 3 | 388 | |||
BC069620.1 | - | 2, 3 | 311 | 7262397 | ||
BC069642.1 | - | 2, 3 | 310 | 7262468 | ||
BC074835.2 | - | 2, 3 | 251 | 30915327 | ||
BC074884.2 | - | 2, 3 | 251 | 30915473 |