Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077356.2 | NP_001070824.1 | 4 | 545 | ||
GenBank mRNA | AB256513.1 | - | 4 | 546 |