Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il36g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113790.1 | NP_001107262.1 | 5 | 469 | ||
XM_008761605.1 | XP_008759827.1 | 5 | 462 | |||
GenBank mRNA | BC158611.1 | - | 5 | 469 | 7384725 |