Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128137.1 | NP_001121609.1 | 1, 2 | 117 | ||
GenBank mRNA | DQ537494.1 | - | 1, 2 | 117 |