Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erich2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024311.1 | NP_001019482.1 | 10, 11 | 1389 | ||
XM_006234340.2 | XP_006234402.1 | 10, 11 | 2495 | |||
XM_008761921.1 | XP_008760143.1 | 3, 4 | 540 | |||
GenBank mRNA | BC082109.1 | - | 10, 11 | 1389 | 7135070 |