Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc30b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127607.1 | NP_001121079.1 | 1 | 653 | ||
GenBank mRNA | BC166739.1 | - | 1 | 653 | 7937387 |
Gene Symbol: Ttc30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271494.1 | NP_001258423.1 | 855 |