Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234700.1 | XP_006234762.1 | 2 | 858 |
Gene Symbol: RGD1561276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_575198.5 | XP_575198.5 | 2 | 951 |