Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234792.2 | XP_006234854.1 | 2 | 242 |
Gene Symbol: RGD1560464 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_343883.6 | XP_343884.5 | 1 | 233 |