Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109204.1 | NP_001102674.1 | 2 | 260 | ||
XM_006234784.2 | XP_006234846.1 | 3 | 438 | |||
GenBank mRNA | BC168875.1 | - | 2 | 224 | 9022295 |