Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109204.1 | NP_001102674.1 | 4 | 543 | ||
XM_006234784.2 | XP_006234846.1 | 5 | 721 | |||
GenBank mRNA | BC168875.1 | - | 4 | 507 | 9022295 |