Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252043.1 | NP_001238972.1 | 9 | 1486 | ||
NM_004783.3 | NP_004774.1 | 9 | 1486 | |||
NM_016151.3 | NP_057235.2 | 9 | 1486 | |||
XM_011545982.1 | XP_011544284.1 | 9 | 1712 | |||
XM_011545983.2 | XP_011544285.1 | 9 | 1712 | |||
XM_011545984.1 | XP_011544286.1 | 9 | 1712 | |||
XM_011545985.1 | XP_011544287.1 | 9 | 1712 | |||
XM_011545986.2 | XP_011544288.1 | 9 | 1712 | |||
GenBank mRNA | AB020688.1 | - | 9 | 1072 | ||
AF061943.1 | - | 9 | 1427 | |||
AF263313.1 | - | 8 | 669 | |||
AK291473.1 | - | 9 | 1003 | |||
AL137701.1 | - | 1 | 1323 | |||
BC015204.1 | - | 9 | 1452 | |||
BC031825.2 | - | 9 | 744 | |||
BC051798.1 | - | 9 | 1222 | |||
BC054495.1 | - | 9 | 1439 | |||
BC136653.1 | - | 9 | 1171 | 9020643 | ||
BC136655.1 | - | 9 | 1171 | 9020645 | ||
BC142663.1 | - | 9 | 826 | 40148112 | ||
BC144344.1 | - | 9 | 1171 | 9052864 | ||
BC151221.1 | - | 9 | 826 | 40148752 | ||
BC152413.1 | - | 9 | 1072 | 8862518 |