Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004784.2 | NP_004775.1 | 6, 7 | 1934 | ||
XM_006714416.3 | XP_006714479.1 | 6, 7 | 1990 | |||
XM_017008839.1 | XP_016864328.1 | 6, 7 | 1902 | |||
XM_017008840.1 | XP_016864329.1 | 6, 7 | 1772 | |||
XM_017008841.1 | XP_016864330.1 | 5, 6 | 3613 | |||
XM_017008842.1 | XP_016864331.1 | 5, 6 | 1217 | |||
XM_017008843.1 | XP_016864332.1 | 3, 4 | 395 | |||
XM_017008844.1 | XP_016864333.1 | 5, 6 | 652 | |||
GenBank mRNA | AF074924.1 | - | 6, 7 | 1934 | ||
AF076605.1 | - | 5, 6 | 1720 | |||
AK295439.1 | - | 4, 5 | 1542 | |||
BC109309.1 | - | 6, 7 | 1781 | 40035856 | ||
BC109310.1 | - | 6, 7 | 1781 | 40035859 |