Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103690020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235962.2 | XP_006236024.1 | 15 | 2268 | ||
XM_006235963.2 | XP_006236025.1 | 1841 | ||||
XM_006235964.2 | XP_006236026.1 | 15 | 2284 |
Gene Symbol: Cd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031561.2 | NP_113749.2 | 1842 | |||
GenBank mRNA | BC072543.1 | - | 1842 | 7105943 |