Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A3R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130012.2 | NP_001123484.1 | 3 | 631 | ||
NM_001252073.1 | NP_001239002.1 | 2 | 248 | |||
NM_001252075.1 | NP_001239004.1 | 2 | 242 | |||
NM_001252076.1 | NP_001239005.1 | 2 | 248 | |||
NM_004785.5 | NP_004776.3 | 3 | 631 | |||
XM_005255702.4 | XP_005255759.1 | 2 | 1242 | |||
XM_006720981.1 | XP_006721044.1 | 2 | 2108 | |||
XM_017023894.1 | XP_016879383.1 | 2 | 1234 | |||
XM_017023895.1 | XP_016879384.1 | 3 | 486 | |||
GenBank mRNA | AF004900.1 | - | 3 | 509 | ||
AF035771.1 | - | 3 | 493 | |||
BC014513.1 | - | 3 | 604 | |||
BC069014.1 | - | 3 | 600 | |||
BC106001.1 | - | 3 | 607 | 5296143 | ||
BQ687174.1 | - | 2 | 126 | |||
BX377861.2 | - | 2 | 234 | |||
CB961565.1 | - | 2 | 269 | |||
U82108.1 | - | 3 | 584 | |||
Z50150.1 | - | 3 | 586 |