Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004786.2 | NP_004777.1 | 3 | 607 | ||
NR_024546.1 | 4 | 712 | ||||
XM_006722580.3 | XP_006722643.1 | 4 | 1212 | |||
XM_011526263.2 | XP_011524565.1 | 5 | 1304 | |||
XM_011526264.2 | XP_011524566.1 | 4 | 451 | |||
XM_017026093.1 | XP_016881582.1 | 5 | 1304 | |||
XM_017026094.1 | XP_016881583.1 | 4 | 451 | |||
GenBank mRNA | AB209263.1 | - | 3 | 345 | ||
AF003938.1 | - | 3 | 563 | |||
AF051896.1 | - | 3 | 488 | |||
AF052659.1 | - | 3 | 507 | |||
AK094926.1 | - | 5 | 1321 | |||
AK313653.1 | - | 3 | 518 | |||
BC001156.1 | - | 3 | 551 | 3534029 | ||
DA248363.1 | - | 4 | 451 | |||
FJ224353.1 | - | 3 | 563 |