Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039344.2 | NP_001034433.1 | 3 | 390 | ||
GenBank mRNA | BC129069.1 | - | 3 | 390 | 7100907 | |
DQ339047.1 | - | 3 | 391 |