Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289135.2 | NP_001276064.1 | 26 | 4716 | ||
NM_001289136.2 | NP_001276065.1 | 26 | 4704 | |||
NM_004787.3 | NP_004778.1 | 27 | 4728 | |||
XM_005248211.3 | XP_005248268.1 | 27 | 4740 | |||
XM_006713986.3 | XP_006714049.1 | 26 | 4668 | |||
XM_011513909.2 | XP_011512211.1 | 27 | 2768 | |||
XM_011513910.2 | XP_011512212.2 | 15 | 3620 | |||
XM_017008845.1 | XP_016864334.1 | 27 | 2755 | |||
GenBank mRNA | AB017168.1 | - | 27 | 3021 | ||
AF055585.1 | - | 26 | 3349 | |||
AF133270.1 | - | 26 | 2805 | |||
AK308444.1 | - | 15 | 1689 | |||
BC117190.1 | - | 27 | 2896 | 40125741 | ||
BC143978.1 | - | 26 | 2872 | 9052496 |