Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109225.1 | NP_001102695.1 | 4 | 1966 | ||
XM_008762812.1 | XP_008761034.1 | 5 | 1986 | |||
XM_008762813.1 | XP_008761035.1 | 4 | 1967 | |||
XM_008762814.1 | XP_008761036.1 | 5 | 2131 | |||
XM_008762815.1 | XP_008761037.1 | 5 | 2207 | |||
XM_008762816.1 | XP_008761038.1 | 5 | 1986 |