Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109226.1 | NP_001102696.1 | 4 | 1011 | ||
XM_006236220.2 | XP_006236282.1 | 4 | 1218 | |||
XM_008762817.1 | XP_008761039.1 | 4 | 902 |