Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271754.1 | NP_001258683.1 | 1 | 308 | ||
NM_001271755.1 | NP_001258684.1 | 1 | 308 | |||
NM_004791.2 | NP_004782.1 | 1 | 308 | |||
XM_005254100.4 | XP_005254157.1 | 1 | 266 | |||
GenBank mRNA | AB008375.1 | - | 1 | 239 | ||
AF072752.1 | - | 1 | 286 | |||
AK291340.1 | - | 1 | 228 | |||
AK297744.1 | - | 1 | 234 | |||
AK308194.1 | - | 1 | 226 | |||
BC036788.1 | - | 1 | 229 | 5753239 | ||
BP284376.1 | - | 2 | 372 | |||
DA578691.1 | - | 1 | 326 |