Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276479.1 | NP_001263408.1 | 2, 3 | 269 | ||
NM_004793.3 | NP_004784.2 | 1, 2 | 554 | |||
NR_076392.1 | 2, 3 | 252 | ||||
XM_011528441.2 | XP_011526743.1 | 1, 2 | 597 | |||
GenBank mRNA | AK096433.1 | - | 2, 3 | 282 | ||
AK096626.1 | - | 2, 3 | 269 | |||
AK127867.1 | - | 2, 3 | 334 | |||
AK298531.1 | - | 2, 3 | 233 | |||
AK309720.1 | - | 2, 3 | 374 | |||
AL096720.1 | - | 1, 2 | 100 | |||
BC000235.1 | - | 1, 2 | 433 | 3350958 | ||
BC109218.1 | - | 1, 2 | 144 | |||
BI906625.1 | - | 1, 2 | 585 | |||
U02389.1 | - | 1, 2 | 530 |