Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ino80b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048233.1 | NP_001041698.1 | 375 | |||
GenBank mRNA | BC105625.1 | - | 375 | 7364756 | ||
FQ214280.1 | - | 762 |
Gene Symbol: NEWGENE_1562258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763087.1 | XP_008761309.1 | 3, 4 | 524 | ||
XM_008763088.1 | XP_008761310.1 | 3, 4 | 524 | |||
XM_008763089.1 | XP_008761311.1 | 3, 4 | 762 |