Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_575584.5 | XP_575584.5 | 3 | 1570 | ||
GenBank mRNA | FQ212435.1 | - | 4 | 2777 |