Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173382.1 | NP_001166853.1 | 7, 8 | 1384 | ||
XM_008763076.1 | XP_008761298.1 | 7, 8 | 1435 |