Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024331.1 | NP_001019502.1 | 3 | 684 | ||
GenBank mRNA | AY593256.1 | - | 3 | 684 |